Methyl-5-fluorobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

99017-1G 99017-500MG 99017-5G 99017-25G
77473-948EA 82.4 USD
77473-948 77486-558 77497-182 77506-066
Methyl-5-fluorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 5-fluorobenzo[b]thiophene-2-carboxylate

Precaución: For research use only

Formula: C₁₀H₇FO₂S
MW: 210.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371535
CAS Number: 154630-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR