1-(4-Bromo-2-fluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

98575-1G 98575-500MG 98575-5G 98575-25G
77473-940EA 415.09 USD
77473-940 77486-552 77497-174 77506-060
1-(4-Bromo-2-fluorophenyl)cyclohexanol ≥95%
1-(4-Bromo-2-fluorophenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₂H₁₄BrFO
MW: 273.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34819576
CAS Number: 2969250-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR