4-(Benzyloxy)-3-bromopyridine ≥95%

Supplier: AOBChem USA

Warning

98068-1G 98068-500MG 98068-5G 98068-25G
77473-936EA 328.57 USD
77473-936 77486-548 77497-170 77506-058
4-(Benzyloxy)-3-bromopyridine ≥95%
4-(Benzyloxy)-3-bromopyridine

Precaución: For research use only

Formula: C₁₂H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD11226996
CAS Number: 1019767-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR