3-Fluoro-6-methoxy-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

97875-1G 97875-500MG 97875-5G
77473-932EA 597.4 USD
77473-932 77486-544 77497-164
3-Fluoro-6-methoxy-2-methylphenol ≥97%
3-Fluoro-6-methoxy-2-methylphenol

Precaución: For research use only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD28515797
CAS Number: 1781641-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR