3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile, 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

97091-1G 97091-500MG 97091-5G 97091-25G
77473-928EA 109.18 USD
77473-928 77486-540 77497-160 77506-050
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester ≥97%
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester

Precaución: For research use only

Formula: C₁₂H₁₃BBrNO₂
Storage Temperature: Ambient
MDL Number: MFCD08669549
CAS Number: 883899-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR