5-Iodo-1-tosyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

96724-1G 96724-500MG 96724-5G 96724-25G
77473-924EA 415.09 USD
77473-924 77486-536 77497-156 77506-046
5-Iodo-1-tosyl-1H-pyrazole ≥97%
5-Iodo-1-tosyl-1H-pyrazole

Precaución: For research use only

Formula: C₁₀H₉IN₂O₂S
MW: 348.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27944675
CAS Number: 142169-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR