1-Bromo-3-chloro-5-ethoxy-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

96665-1G 96665-500MG 96665-5G 96665-25G
77473-920EA 415.09 USD
77473-920 77486-532 77497-152 77506-042
1-Bromo-3-chloro-5-ethoxy-2-fluorobenzene ≥97%
1-Bromo-3-chloro-5-ethoxy-2-fluorobenzene

Precaución: For research use only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34774872
CAS Number: 2385360-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR