5-Chloro-2,4-bis(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

96421-1G 96421-500MG 96421-5G 96421-25G
77473-908EA 415.09 USD
77473-908 77486-520 77497-140 77506-032
5-Chloro-2,4-bis(methylthio)pyridine ≥97%
5-Chloro-2,4-bis(methylthio)pyridine

Precaución: For research use only

Formula: C₇H₈ClNS₂
MW: 205.728 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34819065
CAS Number: 2918950-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR