3-Chloro-4-iodo-2-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

96401-1G 96401-500MG 96401-5G 96401-25G
77473-900EA 415.09 USD
77473-900 77486-512 77497-132 77506-024
3-Chloro-4-iodo-2-(methylthio)pyridine ≥97%
3-Chloro-4-iodo-2-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅ClINS
MW: 285.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774849
CAS Number: 2901106-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR