2-(2-Bromo-3,6-dimethoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

95310-1G 95310-500MG 95310-5G
77473-892EA 537.66 USD
77473-892 77486-504 77497-124
2-(2-Bromo-3,6-dimethoxyphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-3,6-dimethoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₄
MW: 289.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774509
CAS Number: 2918861-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR