1-(2,6-dibromophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

95208-1G 95208-500MG 95208-5G
77473-888EA 820.91 USD
77473-888 77486-500 77497-120
1-(2,6-dibromophenyl)ethanone ≥95%
1-(2,6-dibromophenyl)ethanone

Precaución: For research use only

Formula: C₈H₆Br₂O
MW: 277.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22376086
CAS Number: 99419-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR