2-(5-Fluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

95054-1G 95054-500MG 95054-5G
77473-884EA 685.98 USD
77473-884 77486-496 77497-114
2-(5-Fluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%
2-(5-Fluoro-2-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209222
CAS Number: 773102-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR