3-Iodo-6-isobutoxy-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

94943-1G 94943-500MG 94943-5G 94943-25G
77473-880EA 415.09 USD
77473-880 77486-492 77497-110 77506-014
3-Iodo-6-isobutoxy-2-methylpyridine ≥97%
3-Iodo-6-isobutoxy-2-methylpyridine

Precaución: For research use only

Formula: C₁₀H₁₄INO
MW: 291.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774355
CAS Number: 2901109-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR