6-isobutoxy-2-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

94940-1G 94940-500MG 94940-5G 94940-25G
77473-876EA 397.58 USD
77473-876 77486-488 77497-106 77506-010
6-isobutoxy-2-methylnicotinaldehyde ≥97%
6-isobutoxy-2-methylnicotinaldehyde

Precaución: For research use only

Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD34774353
CAS Number: 2901065-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR