1-(5-Chloro-2-fluoro-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

94730-1G 94730-500MG 94730-5G 94730-25G
77473-872EA 415.09 USD
77473-872 77486-484 77497-102 77506-006
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclobutanol ≥95%
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₂ClFO
MW: 214.664 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774220
CAS Number: 2918947-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR