1-(3-Chloropyridin-4-yl)-2-fluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
94673-1G
94673-500MG
94673-5G
94673-25G
77473-868EA
415.09
USD
77473-868
77486-480
77497-098
77506-002
1-(3-Chloropyridin-4-yl)-2-fluoroethanone ≥95%
1-(3-Chloropyridin-4-yl)-2-fluoroethanone
Precaución: For research use only
Formula:
C₇H₅ClFNO MW: 173.572 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34774168 CAS Number: 2918909-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...