1-(2,6-Difluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

94645-1G 94645-500MG 94645-5G
77473-864EA 497.49 USD
77473-864 77486-476 77497-094
1-(2,6-Difluorophenyl)-2-fluoroethanone ≥95%
1-(2,6-Difluorophenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD34774141
CAS Number: 2294731-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR