Ethyl 2-(6-bromo-2-chloro-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

94617-1G 94617-500MG 94617-5G 94617-25G
77473-860EA 415.09 USD
77473-860 77486-472 77497-090 77505-996
Ethyl 2-(6-bromo-2-chloro-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(6-bromo-2-chloro-3-methylphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₁H₁₀BrClO₃
MW: 305.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774114
CAS Number: 2918869-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR