4-Chloro-5-fluoro-2-iodophenol ≥95%

Supplier: AOBChem USA

Warning

94180-1G 94180-500MG 94180-5G 94180-25G
77473-856EA 246.17 USD
77473-856 77486-468 77497-086 77505-992
4-Chloro-5-fluoro-2-iodophenol ≥95%
4-Chloro-5-fluoro-2-iodophenol

Precaución: For research use only

Formula: C₆H₃ClFIO
MW: 272.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26097249
CAS Number: 1235407-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR