1-chloro-2-fluoro-3-iodo-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

94129-1G 94129-500MG 94129-5G 94129-25G
77473-852EA 415.09 USD
77473-852 77486-464 77497-082 77505-988
1-chloro-2-fluoro-3-iodo-4-(methoxymethoxy)benzene ≥97%
1-chloro-2-fluoro-3-iodo-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇ClFIO₂
MW: 316.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773897
CAS Number: 2901065-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR