2-(2,6-Difluoro-4-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

93984-1G 93984-500MG 93984-5G 93984-25G
77473-848EA 415.09 USD
77473-848 77486-460 77497-078 77505-984
2-(2,6-Difluoro-4-methoxyphenyl)-1,3-dioxolane ≥97%
2-(2,6-Difluoro-4-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773853
CAS Number: 2901106-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR