2,4-Difluoro-3,5-dimethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

93701-1G 93701-500MG 93701-5G 93701-25G
77473-844EA 113.3 USD
77473-844 77486-456 77497-074 77505-980
2,4-Difluoro-3,5-dimethoxybenzoic acid ≥95%
2,4-Difluoro-3,5-dimethoxybenzoic acid

Precaución: For research use only

Formula: C₉H₈F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09839225
CAS Number: 1003709-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR