1-Bromo-2-chloro-3-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

93381-1G 93381-500MG 93381-5G
77473-836EA 920.82 USD
77473-836 77486-448 77497-066
1-Bromo-2-chloro-3-ethylbenzene ≥95%
1-Bromo-2-chloro-3-ethylbenzene

Precaución: For research use only

Formula: C₈H₈BrCl
Storage Temperature: Ambient
MDL Number: MFCD22490801
CAS Number: 1369869-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR