2-Fluoro-1-(4-Fluoro-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

93169-1G 93169-500MG 93169-5G
77473-832EA 553.11 USD
77473-832 77486-444 77497-062
2-Fluoro-1-(4-Fluoro-2-methylphenyl)ethanone ≥95%
2-Fluoro-1-(4-Fluoro-2-methylphenyl)ethanone

Precaución: For research use only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773442
CAS Number: 2171961-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR