1-(4-propylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93130-1G 93130-500MG 93130-5G
77473-828EA 546.93 USD
77473-828 77486-440 77497-058
1-(4-propylphenyl)cyclobutanol ≥95%
1-(4-propylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD21788619
CAS Number: 1532792-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR