tert-Butyl 2-fluoro-3-methoxy-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

92911-1G 92911-500MG 92911-5G 92911-25G
77473-820EA 415.09 USD
77473-820 77486-432 77497-050 77505-968
tert-Butyl 2-fluoro-3-methoxy-5-methylbenzoate ≥95%
tert-Butyl 2-fluoro-3-methoxy-5-methylbenzoate

Precaución: For research use only

Formula: C₁₃H₁₇FO₃
MW: 240.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773321
CAS Number: 2918892-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR