2-(2,4-Dichlorophenyl)-2-oxoacetic acid ≥95%

Supplier: AOBChem USA

Warning

92577-1G 92577-500MG 92577-5G 92577-25G
77473-816EA 297.67 USD
77473-816 77486-428 77497-046 77505-966
2-(2,4-Dichlorophenyl)-2-oxoacetic acid ≥95%
2-(2,4-Dichlorophenyl)-2-oxoacetic acid

Precaución: For research use only

Formula: C₈H₄Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD14610209
CAS Number: 32375-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR