2-(3-Bromophenyl)-2-oxoacetic acid ≥95%

Supplier: AOBChem USA

Warning

92493-1G 92493-500MG 92493-5G 92493-25G
77473-812EA 89.61 USD
77473-812 77486-424 77497-042 77505-962
2-(3-Bromophenyl)-2-oxoacetic acid ≥95%
2-(3-Bromophenyl)-2-oxoacetic acid

Precaución: For research use only

Formula: C₈H₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD07698694
CAS Number: 7194-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR