1-(5-Chloro-2-methoxy-4-(methoxymethoxy)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

92484-1G 92484-500MG 92484-5G 92484-25G
77473-808EA 415.09 USD
77473-808 77486-420 77497-038 77505-958
1-(5-Chloro-2-methoxy-4-(methoxymethoxy)phenyl)ethanone ≥97%
1-(5-Chloro-2-methoxy-4-(methoxymethoxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₃ClO₄
MW: 244.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773208
CAS Number: 2901105-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR