(3-Bromophenyl)(5-Bromopyridin-2-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

92332-1G 92332-500MG 92332-5G 92332-25G
77473-804EA 226.6 USD
77473-804 77486-416 77497-034 77505-954
(3-Bromophenyl)(5-Bromopyridin-2-yl)methanone ≥97%
(3-Bromophenyl)(5-Bromopyridin-2-yl)methanone

Precaución: For research use only

Formula: C₁₂H₇Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD34818285
CAS Number: 2001519-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR