(3,5-Diethylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

92191-1G 92191-500MG 92191-5G
77473-796EA 727.18 USD
77473-796 77486-408 77497-026
(3,5-Diethylphenyl)boronic acid ≥95%
(3,5-Diethylphenyl)boronic acid

Precaución: For research use only

Formula: C₁₀H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD30166787
CAS Number: 1147894-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR