1-Bromo-2-ethyl-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

92063-1G 92063-500MG 92063-5G
77473-792EA 715.85 USD
77473-792 77486-404 77497-022
1-Bromo-2-ethyl-3-fluorobenzene ≥97%
1-Bromo-2-ethyl-3-fluorobenzene

Precaución: For research use only

Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD22490760
CAS Number: 1780931-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR