4-Bromo-3-ethylaniline ≥95%

Supplier: AOBChem USA

Warning

91908-1G 91908-500MG 91908-5G 91908-25G
77473-788EA 106.09 USD
77473-788 77486-400 77497-018 77505-946
4-Bromo-3-ethylaniline ≥95%
4-Bromo-3-ethylaniline

Precaución: For research use only

Formula: C₈H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD19381654
CAS Number: 52121-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR