2-(2,4-Dichloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

91860-1G 91860-500MG 91860-5G 91860-25G
77473-784EA 415.09 USD
77473-784 77486-396 77497-014 77505-942
2-(2,4-Dichloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥95%
2-(2,4-Dichloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₂Cl₂O₂S
MW: 279.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772941
CAS Number: 2901065-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR