2-Bromo-6-ethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

91736-1G 91736-500MG 91736-5G 91736-25G
77473-780EA 389.34 USD
77473-780 77486-392 77497-010 77505-938
2-Bromo-6-ethoxyphenol ≥97%
2-Bromo-6-ethoxyphenol

Precaución: For research use only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22414722
CAS Number: 187961-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR