2-Bromo-4-methoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

91596-1G 91596-500MG 91596-5G 91596-25G
77473-776EA 127.72 USD
77473-776 77486-388 77497-006 77505-934
2-Bromo-4-methoxy-1,3-dimethylbenzene ≥97%
2-Bromo-4-methoxy-1,3-dimethylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28401429
CAS Number: 23453-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR