1-(Benzofuran-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

91402-1G 91402-500MG 91402-5G 91402-25G
77473-772EA 255.44 USD
77473-772 77486-384 77497-002 77505-930
1-(Benzofuran-2-yl)cyclobutanol ≥95%
1-(Benzofuran-2-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34818174
CAS Number: 1601939-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR