Ethyl-5-methylthiophene-2-glyoxylate ≥95%

Supplier: AOBChem USA

Warning

91389-1G 91389-500MG 91389-5G 91389-25G
77473-768EA 284.28 USD
77473-768 77486-380 77496-998 77505-926
Ethyl-5-methylthiophene-2-glyoxylate ≥95%
Ethyl-5-methylthiophene-2-glyoxylate

Precaución: For research use only

Formula: C₉H₁₀O₃S
Storage Temperature: Ambient
MDL Number: MFCD07782947
CAS Number: 50845-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR