2-Bromo-1-ethoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

90771-1G 90771-500MG 90771-5G 90771-25G
77473-760EA 232.78 USD
77473-760 77486-372 77496-990 77505-920
2-Bromo-1-ethoxy-3-methylbenzene ≥95%
2-Bromo-1-ethoxy-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738586
CAS Number: 1805027-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR