4-Bromo-2,6-diisopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

90618-1G 90618-500MG 90618-5G
77473-756EA 1233.94 USD
77473-756 77486-368 77496-986
4-Bromo-2,6-diisopropylbenzoic acid ≥97%
4-Bromo-2,6-diisopropylbenzoic acid

Precaución: For research use only

Formula: C₁₃H₁₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD18917126
CAS Number: 1349718-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR