2,4-Dibromo-5-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

90549-1G 90549-500MG 90549-5G 90549-25G
77473-752EA 109.18 USD
77473-752 77486-364 77496-982 77505-916
2,4-Dibromo-5-methoxypyridine ≥95%
2,4-Dibromo-5-methoxypyridine

Precaución: For research use only

Formula: C₆H₅Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD18257700
CAS Number: 84539-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR