4-Bromo-2,6-difluoro-3-methylphenol ≥95%

Supplier: AOBChem USA

Warning

90236-1G 90236-500MG 90236-5G 90236-25G
77473-748EA 120.51 USD
77473-748 77486-360 77496-978 77505-912
4-Bromo-2,6-difluoro-3-methylphenol ≥95%
4-Bromo-2,6-difluoro-3-methylphenol

Precaución: For research use only

Formula: C₇H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD21609557
CAS Number: 1365272-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR