Methyl 5-(benzyloxy)picolinate ≥95%

Supplier: AOBChem USA

Warning

90123-1G 90123-500MG 90123-5G 90123-25G
77473-744EA 287.37 USD
77473-744 77486-356 77496-972 77505-906
Methyl 5-(benzyloxy)picolinate ≥95%
Methyl 5-(benzyloxy)picolinate

Precaución: For research use only

Formula: C₁₄H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD32704786
CAS Number: 74386-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR