1-(3-Chloro-2-fluorophenyl)cyclopentanol ≥95%
Supplier: AOBChem USA
|
Warning
|
90061-1G
90061-500MG
90061-5G
90061-25G
77473-740EA
415.09
USD
77473-740
77486-352
77496-968
77505-902
1-(3-Chloro-2-fluorophenyl)cyclopentanol ≥95%
1-(3-Chloro-2-fluorophenyl)cyclopentanol
Precaución: For research use only
Formula:
C₁₁H₁₂ClFO MW: 214.664 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34772121 CAS Number: 2918900-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...