2-Chloro-6-fluoronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

90010-1G 90010-500MG 90010-5G
77473-736EA 529.42 USD
77473-736 77486-348 77496-964
2-Chloro-6-fluoronicotinic acid ≥95%
2-Chloro-6-fluoronicotinic acid

Precaución: For research use only

Formula: C₆H₃ClFNO₂
MW: 175.545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185820
CAS Number: 1211533-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR