5-(cyclopropylmethoxy)picolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

89903-1G 89903-500MG 89903-5G
77473-732EA 518.09 USD
77473-732 77486-344 77496-960
5-(cyclopropylmethoxy)picolinaldehyde ≥97%
5-(cyclopropylmethoxy)picolinaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23769793
CAS Number: 1502637-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR