2-Iodo-5-(methoxymethyl)thiophene ≥97%

Supplier: AOBChem USA

Warning

89888-1G 89888-500MG 89888-5G
77473-728EA 546.93 USD
77473-728 77486-340 77496-956
2-Iodo-5-(methoxymethyl)thiophene ≥97%
2-Iodo-5-(methoxymethyl)thiophene

Precaución: For research use only

Formula: C₆H₇IOS
MW: 254.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28670235
CAS Number: 90724-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR