2,2,2-trifluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

89841-1G 89841-500MG 89841-5G 89841-25G
77473-724EA 415.09 USD
77473-724 77486-336 77496-952 77505-892
2,2,2-trifluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(3-methyl-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34818013
CAS Number: 2921797-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR