1-(5-Methoxy-2-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

89627-1G 89627-500MG 89627-5G 89627-25G
77473-716EA 226.6 USD
77473-716 77486-328 77496-944 77505-886
1-(5-Methoxy-2-methylphenyl)ethanol ≥97%
1-(5-Methoxy-2-methylphenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD27931762
CAS Number: 89691-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR