1-(4-Isopropylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89172-1G 89172-500MG 89172-5G 89172-25G
77473-704EA 258.53 USD
77473-704 77486-316 77496-932 77505-878
1-(4-Isopropylphenyl)cyclohexanol ≥95%
1-(4-Isopropylphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD17223735
CAS Number: 37920-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR